2,3-Dichlorisonicotinsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,3-Dichloropyridine-4-carboxylic acid
H63733.06
H63733.06EA
106
EUR
H63733.06
2,3-Dichlorisonicotinsäure ≥97%
2,3-Dichlorisonicotinsäure
Purity: 97%
CAS No.: 184416-84-0
Molecular Formula: C6H3Cl2NO2
Formula Weight: 192.00
Physical Form: Solid
Melting Point: 203-207°
MDL No.: MFCD01861977
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol Schmelzpunkt: 203…207 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861977 CAS-Nummer: 184416-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...