1-Acetyl-5-bromoindole 97%

Lieferant: Thermo Scientific

H63745.03
H63745.03EA 111 EUR
H63745.03
1-Acetyl-5-bromoindole 97%
1-Acetyl-5-bromoindole

Purity: 97% CAS No.: 61995-52-6 Molecular Formula: C10H8BrNO Formula Weight: 238.08 Physical Form: Solid Beilstein Reference: 1283925 MDL No.: MFCD00238530

Formel: C₁₀H₈BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00238530
CAS-Nummer: 61995-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR