Methyl-6-(hydroxymethyl)picolinat 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 6-(hydroxymethyl)picolinate, Methyl 6-(hydroxymethyl)pyridine-2-carboxylate
H63793.03
H63793.03EA
81.5
EUR
H63793.03
Methyl-6-(hydroxymethyl)picolinat 95%
Methyl-6-(hydroxymethyl)picolinat
Purity: 95%
CAS No.: 39977-44-1
Molecular Formula: C8H9NO3
Formula Weight: 167.16
Physical Form: Solid
Appearance: White
MDL No.: MFCD03840850
Formel:
C₈H₉NO₃ Molecular Weight: 167,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840850 CAS-Nummer: 39977-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...