1-(3-Chlor-4-methoxyphenyl)methanaminhydrochlorid 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Chloro-4-methoxybenzylamine hydrochloride
H63806.06
H63806.06EA
123
EUR
H63806.06
1-(3-Chlor-4-methoxyphenyl)methanaminhydrochlorid 97%
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride
Purity: 97%
CAS No.: 41965-95-1
Molecular Formula: C8H10ClNO.HCl
Formula Weight: 208.09
Physical Form: Solid
Melting Point: 250-255°
UN No.: UN2811
MDL No.: MFCD01211460
Formel:
C₈H₁₁Cl₂NO Molecular Weight: 208.09 g/mol Schmelzpunkt: 250…255 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01211460 CAS-Nummer: 41965-95-1 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...