8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%

Lieferant: Thermo Fisher Scientific
Achtung

H63825.MD H63825.03 H63825.06
H63825.MDEA 68.1 EUR
H63825.MD H63825.03 H63825.06
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride

Purity: 97% CAS No.: 54745-74-3 Molecular Formula: C6H11NO.HCl Formula Weight: 149.62 Physical Form: Solid MDL No.: MFCD09800611

Formel: C₆H₁₂ClNO
Molecular Weight: 149.62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800611
CAS-Nummer: 54745-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR