8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H63825.MD
H63825.03
H63825.06
H63825.MDEA
68.1
EUR
H63825.MD
H63825.03
H63825.06
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride 97%
8-Oxa-3-azabicyclo[3.2.1]octane hydrochloride
Purity: 97%
CAS No.: 54745-74-3
Molecular Formula: C6H11NO.HCl
Formula Weight: 149.62
Physical Form: Solid
MDL No.: MFCD09800611
Formel:
C₆H₁₂ClNO Molecular Weight: 149.62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800611 CAS-Nummer: 54745-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...