2-Iodo-5-trifluoromethylpyridine 96%

Lieferant: Thermo Fisher Scientific
Achtung

H63848.03 H63848.06
H63848.03EA 116 EUR
H63848.03 H63848.06
2-Iodo-5-trifluoromethylpyridine 96%
2-Iodo-5-trifluoromethylpyridine

Purity: 96% CAS No.: 100366-75-4 Molecular Formula: C6H3F3IN Formula Weight: 272.99 MDL No.: MFCD08741351

Formel: C₆H₃F₃IN
Lagertemperatur: Raumtemperatur
MDL: MFCD08741351
CAS-Nummer: 100366-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR