1-(Methylsulfonyl)piperazin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H63870.06
H63870.14
H63870.22
H63870.06EA
47.5
EUR
H63870.06
H63870.14
H63870.22
1-(Methylsulfonyl)piperazin 97%
1-(Methylsulfonyl)piperazin
Purity: 97%
CAS No.: 55276-43-2
Molecular Formula: C5H12N2O2S
Formula Weight: 164.23
Physical Form: Solid
MDL No.: MFCD02153225
Formel:
C₅H₁₂N₂O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD02153225 CAS-Nummer: 55276-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...