Ac-Glu(OtBu)-OH 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Acetyl-L-glutamic acid 5-tert-butyl ester
H63871.03
H63871.06
H63871.03EA
81.7
EUR
H63871.03
H63871.06
Ac-Glu(OtBu)-OH 95%
Ac-Glu(OtBu)-OH
Purity: 95%
CAS No.: 84192-88-1
Molecular Formula: C11H19NO
Formula Weight: 245.27
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00236754
Formel:
C₁₁H₁₉NO₅ |
MDL:
MFCD00236754 CAS-Nummer: 84192-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...