2-Bromo-4,5-difluorophenylacetic acid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H63902.03
H63902.06
H63902.03EA
83.9
EUR
H63902.03
H63902.06
2-Bromo-4,5-difluorophenylacetic acid 98%
2-Bromo-4,5-difluorophenylacetic acid
Purity: 98%
CAS No.: 883502-07-6
Molecular Formula: C8H5BrF2O2
Formula Weight: 251.03
Physical Form: Solid
Appearance: Off-white
MDL No.: MFCD04973423
Formel:
C₈H₅BrF₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973423 CAS-Nummer: 883502-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...