5-(Trifluoromethyl)pyridine-3-carboxylic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-(Trifluoromethyl)nicotinic acid
H63906.03
H63906.03EA
122
EUR
H63906.03
5-(Trifluoromethyl)pyridine-3-carboxylic acid 97%
5-(Trifluoromethyl)pyridine-3-carboxylic acid
Purity: 97%
CAS No.: 131747-40-5
Molecular Formula: C7H4F3NO2
Formula Weight: 191.11
Physical Form: Solid
Appearance: Off-white
Melting Point: 184-189°
UN No.: UN2811
MDL No.: MFCD07774128
Formel:
C₇H₄F₃NO₂ Molecular Weight: 191.11 g/mol Schmelzpunkt: 184…189 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07774128 CAS-Nummer: 131747-40-5 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...