6-Fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Fluoro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole

H63920.03
H63920.03EA 220 EUR
H63920.03
6-Fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole 97%
6-Fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole

Purity: 97% CAS No.: 180161-14-2 Molecular Formula: C13H13FN2 Formula Weight: 216.25 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Yellow to brown Beilstein Reference: 8627107 MDL No.: MFCD04114772

Formel: C₁₃H₁₃FN₂
MDL: MFCD04114772
CAS-Nummer: 180161-14-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR