5-Nitrogramine 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Dimethylaminomethyl)-5-nitroindole
H63939.03
H63939.03EA
220
EUR
H63939.03
5-Nitrogramine 95%
5-Nitrogramine
Purity: 95%
CAS No.: 3414-64-0
Molecular Formula: C11H13N3O2
Formula Weight: 219.24
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 216692
MDL No.: MFCD00544197
Formel:
C₁₁H₁₃N₃O₂ |
MDL:
MFCD00544197 CAS-Nummer: 3414-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...