3-Bromo-7-nitroindole 97%

Lieferant: Thermo Scientific
Achtung

H63940.06
H63940.06EA 154 EUR
H63940.06
3-Bromo-7-nitroindole 97%
3-Bromo-7-nitroindole

Purity: 97% CAS No.: 397864-11-8 Molecular Formula: C8H5BrN2O2 Formula Weight: 241.04 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD02684155

Formel: C₈H₅BrN₂O₂
MDL: MFCD02684155
CAS-Nummer: 397864-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR