7-Bromchinolin 95%

Lieferant: Thermo Fisher Scientific
Achtung

H63987.03 H63987.06
H63987.03EA 139 EUR
H63987.03 H63987.06
7-Bromchinolin 95%
7-Bromoquinoline

Purity: 95% CAS No.: 4965-36-0 Molecular Formula: C9H6BrN Formula Weight: 208.06 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD03695823

Formel: C₉H₆BrN
Lagertemperatur: Gefrierschrank
MDL: MFCD03695823
CAS-Nummer: 4965-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR