Fmoc-N-Me-Nle-OH 96%

Lieferant: Thermo Fisher Scientific

H63992.06
H63992.06EA 75.5 EUR
H63992.06
Fmoc-N-Me-Nle-OH 96%
Fmoc-N-Me-Nle-OH

Purity: 96% CAS No.: 112883-42-8 Molecular Formula: C22H25NO4 Formula Weight: 367.44 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD00235878

Formel: C₂₂H₂₅NO₄
Lagertemperatur: Kühlschrank
MDL: MFCD00235878
CAS-Nummer: 112883-42-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR