(R)-Morpholin-2-ylmethanol hydrochloride 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(R)-2-Hydroxymethylmorpholine hydrochloride, (R)-2-Morpholinemethanol hydrochloride
H64003.03
H64003.03EA
263
EUR
H64003.03
(R)-Morpholin-2-ylmethanol hydrochloride 95%
(R)-Morpholin-2-ylmethanol hydrochloride
Purity: 95%
CAS No.: 1436436-17-7
Molecular Formula: C5H11NO2.HCl
Formula Weight: 153.61
Physical Form: Solid
MDL No.: MFCD11052541
Formel:
C₅H₁₁NO₂·HCl Molecular Weight: 153,61 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11052541 CAS-Nummer: 1436436-17-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...