3-Methyl-3-(p-toluenesulphonyloxymethyl)oxetane 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64011.03
H64011.06
H64011.03EA
114
EUR
H64011.03
H64011.06
3-Methyl-3-(p-toluenesulphonyloxymethyl)oxetane 98%
3-Methyl-3-(p-toluenesulphonyloxymethyl)oxetane
Purity: 98%
CAS No.: 99314-44-0
Molecular Formula: C12H16O4S
Formula Weight: 256.32
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
Melting Point: 50-51°
Beilstein Reference: 4441465
MDL No.: MFCD08275394
Formel:
C₁₂H₁₆O₄S Schmelzpunkt: 50…51 °C |
MDL:
MFCD08275394 CAS-Nummer: 99314-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...