Methyl-6-chlorpicolinat 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 6-chloropyridine-2-carboxylate, Methyl 6-chloropicolinate

H64023.06 H64023.14
H64023.06EA 47.8 EUR
H64023.06 H64023.14
Methyl-6-chlorpicolinat 95%
Methyl-6-chlorpicolinat

Purity: 95% CAS No.: 6636-55-1 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Pale yellow Melting Point: 92-98° lit. Beilstein Reference: 128580 MDL No.: MFCD00155391

Formel: C₇H₆ClNO₂
Molecular Weight: 171.58 g/mol
Schmelzpunkt: 92…98 °C (Lit.)
MDL: MFCD00155391
CAS-Nummer: 6636-55-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR