6-Chloro-5-methoxypyridin-3-amine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Amino-6-chloro-5-methoxypyridine

H64033.03
H64033.03EA 449 EUR
H64033.03
6-Chloro-5-methoxypyridin-3-amine 97%
6-Chloro-5-methoxypyridin-3-amine

Purity: 97% CAS No.: 75711-01-2 Molecular Formula: C6H7ClN2O Formula Weight: 158.59 Physical Form: Solid Beilstein Reference: 15026636 MDL No.: MFCD09952112

Formel: C₆H₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09952112
CAS-Nummer: 75711-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR