5-Bromopyrimidine-2-carboxylic acid 97%

Lieferant: Thermo Scientific
Achtung

H64041.06
H64041.06EA 272 EUR
H64041.06
5-Bromopyrimidine-2-carboxylic acid 97%
5-Bromopyrimidine-2-carboxylic acid

Purity: 98% CAS No.: 37131-87-6 Molecular Formula: C5H3BrN2O2 Formula Weight: 203.00 Physical Form: Solid Melting Point: 193-198° Beilstein Reference: 121432 MDL No.: MFCD00496793

Formel: C₅H₃BrN₂O₂
Molecular Weight: 203,00 g/mol
Schmelzpunkt: 193…198 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00496793
CAS-Nummer: 37131-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR