2-Chloropyrimidine-5-boronic acid pinacol ester 98%

Lieferant: Thermo Fisher Scientific
Achtung

H64052.06
H64052.06EA 591 EUR
H64052.06
2-Chloropyrimidine-5-boronic acid pinacol ester 98%
2-Chloropyrimidine-5-boronic acid pinacol ester

Purity: 98% CAS No.: 1003845-08-6 Molecular Formula: C10H14BClN2O2 Formula Weight: 240.49 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 15622160 MDL No.: MFCD11856048

Formel: C₁₀H₁₄BClN₂O₂
MDL: MFCD11856048
CAS-Nummer: 1003845-08-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR