Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64073.14
H64073.14EA
595
EUR
H64073.14
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate 95%
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate
Purity: 95%
CAS No.: 51145-57-4
Molecular Formula: C9H15NO3
Formula Weight: 185.22
Physical Form: Liquid
Appearance: Pale red
MDL No.: MFCD00087563
Formel:
C₉H₁₅NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00087563 CAS-Nummer: 51145-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...