2-[3-(Trifluormethyl)phenyl]ethanamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-(Trifluoromethyl)phenethylamine

H64084.03 H64084.06 H64084.14
H64084.03EA 42.3 EUR
H64084.03 H64084.06 H64084.14
2-[3-(Trifluormethyl)phenyl]ethanamin 98%
2-(3-Trifluoromethylphenyl)ethylamine

Purity: 98% CAS No.: 52516-30-0 Molecular Formula: C9H10F3N Formula Weight: 189.18 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.18 Refractive Index: 1.4645 Sensitivity: Light Sensitive Appearance: Colourless to pale yellow Boiling Point: 113°/22mm Beilstein Reference: 4398117 UN No.: UN2735 MDL No.: MFCD00040756

Formel: C₉H₁₀F₃N
Molecular Weight: 189.18 g/mol
Siedepunkt: 113 °C (22 mmHg)
Dichte: 1.18
MDL: MFCD00040756
CAS-Nummer: 52516-30-0
EINECS: 257-988-3
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR