1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-tert-Butyl 4-ethyl 3-oxopiperidine-1,4-dicarboxylate, Ethyl 1-Boc-3-oxopiperidine-4-carboxylate
H64102.06
H64102.06EA
188
EUR
H64102.06
1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate 97%
1-tert-Butyl-4-ethyl-3-oxopiperidine-1,4-dicarboxylate
Purity: 97%
CAS No.: 71233-25-5
Molecular Formula: C13H21NO5
Formula Weight: 271.31
Physical Form: Liquid
Beilstein Reference: 421506
MDL No.: MFCD09878815
Formel:
C₁₃H₂₁NO₅ Molecular Weight: 271,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878815 CAS-Nummer: 71233-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...