2-Brom-4'-(trifluormethoxy)acetophenon 97%

Lieferant: Thermo Fisher Scientific
Achtung

H64105.03 H64105.06
H64105.03EA 76.9 EUR
H64105.03 H64105.06
2-Brom-4'-(trifluormethoxy)acetophenon 97%
2-Brom-4'-(trifluormethoxy)acetophenon

Purity: 97% CAS No.: 103962-10-3 Molecular Formula: C9H6BrF3O2 Formula Weight: 283.04 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Solid Appearance: Pale yellow Melting Point: 48-55° Beilstein Reference: 7479010 MDL No.: MFCD00052333

Formel: C₉H₆BrF₃O₂
Schmelzpunkt: 48…55 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00052333
CAS-Nummer: 103962-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR