2-(Trifluormethyl)-5-pyridinboronsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-(Trifluoromethyl)-3-pyridylboronic acid, 2-(Trifluoromethyl)pyridin-5-ylboronic acid, 2-(Trifluoromethyl)pyridine-5-boronic acid, (6-(Trifluoromethyl)pyridin-3-yl)boronic acid
H64106.03
H64106.06
H64106.03EA
116
EUR
H64106.03
H64106.06
2-(Trifluormethyl)-5-pyridinboronsäure 98%
2-(Trifluormethyl)-5-pyridinboronsäure
Purity: 98%
CAS No.: 868662-36-6
Molecular Formula: C6H5BF3NO2
Formula Weight: 190.92
Physical Form: Solid
MDL No.: MFCD07375381
Formel:
C₆H₅BF₃NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375381 CAS-Nummer: 868662-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...