α,α,α,2,5-Pentafluortoluol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,5-Difluorobenzotrifluoride
H64111.03
H64111.06
H64111.03EA
44.7
EUR
H64111.03
H64111.06
α,α,α,2,5-Pentafluortoluol 98%
α,α,α,2,5-Pentafluortoluol
Purity: 98%
CAS No.: 393-38-4
Molecular Formula: C7H3F5
Formula Weight: 182.09
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.424
Refractive Index: 1.3940
Appearance: Colourless to pale yellow
Melting Point: -14°
Boiling Point: 107-108°
Beilstein Reference: 2049494
MDL No.: MFCD00042494
Formel:
C₇H₃F₅ Molecular Weight: 182.09 g/mol Siedepunkt: 107…108 °C Schmelzpunkt: –14 °C Dichte: 1.424 |
MDL:
MFCD00042494 CAS-Nummer: 393-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...