1-(Brommethyl)naphthalin 98%

Lieferant: Thermo Scientific
Gefahr

H64116.14
H64116.14EA 109 EUR
H64116.14
1-(Brommethyl)naphthalin 98%
1-(Bromomethyl)naphthalene

Purity: 98% CAS No.: 3163-27-7 Molecular Formula: C11H9Br Formula Weight: 221.10 Storage Temperature: 2°C to 8°C Physical Form: Powder Density: 1.44 Refractive Index: 1.6600 Appearance: Off-white Boiling Point: 213°/100mm Beilstein Reference: 742417 UN No.: UN3261 MDL No.: MFCD00010804

Formel: C₁₁H₉Br
Molecular Weight: 221,10 g/mol
Siedepunkt: 213 °C (100 mmHg)
Dichte: 1,44
MDL: MFCD00010804
CAS-Nummer: 3163-27-7
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR