3-Amino-1-adamantanol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Aminoadamantan-1-ol

H64137.14
H64137.14EA 259 EUR
H64137.14
3-Amino-1-adamantanol ≥98%
3-Amino-1-adamantanol

Purity: 98+% CAS No.: 702-82-9 Molecular Formula: C10H17NO Formula Weight: 167.25 Flash Point: 110°(230°F) Physical Form: Solid Melting Point: 265° lit. Beilstein Reference: 2802127 MDL No.: MFCD01821204

Formel: C₁₀H₁₇NO
Molecular Weight: 167,25 g/mol
Schmelzpunkt: 265 °C (Lit.)
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD01821204
CAS-Nummer: 702-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR