Cyclohexylboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
H64178.03
H64178.06
H64178.14
H64178.03EA
45.3
EUR
H64178.03
H64178.06
H64178.14
Cyclohexylboronsäurepinakolester 97%
Cyclohexylboronsäurepinakolester
Purity: 97%
CAS No.: 87100-15-0
Molecular Formula: C12H23BO2
Formula Weight: 210.12
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD04038749
Formel:
C₁₂H₂₃BO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038749 CAS-Nummer: 87100-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...