4-(Trifluormethyl)anisol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98%
CAS No.: 402-52-8
Molecular Formula: C8H7F3O
Formula Weight: 176.14
Storage Temperature: 2°C to 8°C
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 1.245
Refractive Index: 1.4450
Appearance: Colourless to pale yellow
Melting Point: -9°
Boiling Point: 169°/754mm
UN No.: UN1993
MDL No.: MFCD03094456
Formel:
C₈H₇F₃O Siedepunkt: 169 °C (754 mmHg) Schmelzpunkt: –9 °C Dichte: 1,245 Flammpunkt: 60 °C (140 °F) |
MDL:
MFCD03094456 CAS-Nummer: 402-52-8 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...