5-Brom-2-hydroxy-3-picolin 98%

Lieferant: Thermo Fisher Scientific
Achtung

H64253.22
H64253.22EA 298 EUR
H64253.22
5-Brom-2-hydroxy-3-picolin 98%
5-Brom-2-hydroxy-3-picolin

Purity: 98% CAS No.: 89488-30-2 Molecular Formula: C6H6BrNO Formula Weight: 188.02 Physical Form: Solid Appearance: Off-white Melting Point: 170-174° lit. Beilstein Reference: 1447443 MDL No.: MFCD03427657

Formel: C₆H₆BrNO
Schmelzpunkt: 170…174 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD03427657
CAS-Nummer: 89488-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR