Methyl-3-(4-hydroxyphenyl)propanoat 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-(4-Hydroxyphenyl)propionic Acid Methyl Ester, Methyl 3-(4-hydroxyphenyl)propanoate, Methyl 3-(4-hydroxyphenyl)propionate, 3-(4-Hydroxy-phenyl)-propionic acid-OMe

H64255.14 H64255.22
H64255.14EA 102 EUR
H64255.14 H64255.22
Methyl-3-(4-hydroxyphenyl)propanoat 98%
Methyl-3-(4-hydroxyphenyl)propionate

Purity: 98+% CAS No.: 5597-50-2 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 113°(235°F) Physical Form: Powder Appearance: Off-white Melting Point: 39-41° lit. Boiling Point: 108°/11mm Beilstein Reference: 2211073 MDL No.: MFCD00071577

Formel: C₁₀H₁₂O₃
Siedepunkt: 108 °C (11 mmHg)
Schmelzpunkt: 39…41 °C (Lit.)
Flammpunkt: >113 °C (235 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00071577
CAS-Nummer: 5597-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR