3-Cyano-4-methoxy-2-pyridone 95%

Lieferant: Thermo Fisher Scientific
Achtung

H64279.14
H64279.14EA 370 EUR
H64279.14
3-Cyano-4-methoxy-2-pyridone 95%
3-Cyano-4-methoxy-2-pyridone

Purity: 95% CAS No.: 21642-98-8 Molecular Formula: C7H6N2O2 Formula Weight: 150.14 Physical Form: Crystalline solid Sensitivity: Hygroscopic Appearance: Pale yellow Melting Point: 278° Beilstein Reference: 5929290 UN No.: UN3439 MDL No.: MFCD00975442

Formel: C₇H₆N₂O₂
Schmelzpunkt: 278 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08059559
CAS-Nummer: 21642-98-8
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR