Methyl-2-amino-5-(trifluoromethyl)benzoate 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 2-amino-5-(trifluoromethyl)benzoate

H64283.03 H64283.06
H64283.03EA 93 EUR
H64283.03 H64283.06
Methyl-2-amino-5-(trifluoromethyl)benzoate 98%
Methyl-2-amino-5-(trifluoromethyl)benzoate

Purity: 98% CAS No.: 117324-58-0 Molecular Formula: C9H8F3NO2 Formula Weight: 219.16 Physical Form: Solid Appearance: White MDL No.: MFCD08234902

Formel: C₉H₈F₃NO₂
Molecular Weight: 219.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234902
CAS-Nummer: 117324-58-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR