2-Bromo-3-fluoroisonicotinic acid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Bromo-3-fluoropyridine-4-carboxylic acid

H64290.06
H64290.06EA 241 EUR
H64290.06
2-Bromo-3-fluoroisonicotinic acid 98%
2-Bromo-3-fluoroisonicotinic acid

Purity: 98% CAS No.: 1211530-89-0 Molecular Formula: C6H3BrFNO2 Formula Weight: 220.00 Physical Form: Solid MDL No.: MFCD13185796

Formel: C₆H₃BrFNO₂
Molecular Weight: 219.997 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185796
CAS-Nummer: 1211530-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR