4-Methoxy-2-methylbenzaldehyd 95%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Methyl-4-methoxybenzaldehyde

H64295.03 H64295.06
H64295.03EA 46.4 EUR
H64295.03 H64295.06
4-Methoxy-2-methylbenzaldehyd 95%
4-Methoxy-2-methylbenzaldehyde

Purity: 95% CAS No.: 52289-54-0 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.107 Refractive Index: 1.5710 Appearance: Pale yellow Boiling Point: 262-267° lit. Beilstein Reference: 1933786 MDL No.: MFCD02261771

Formel: C₉H₁₀O₂
Siedepunkt: 262…267 °C (Lit.)
Dichte: 1,107
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD02261771
CAS-Nummer: 52289-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR