2-Brom-3-(trifluormethyl)anilin 98%

Lieferant: Thermo Scientific
Achtung

H64309.06
H64309.06EA 109 EUR
H64309.06
2-Brom-3-(trifluormethyl)anilin 98%
2-Brom-3-(trifluormethyl)anilin

Purity: 98% CAS No.: 58458-10-9 Molecular Formula: C7H5BrF3N Formula Weight: 240.02 Physical Form: Solid Appearance: Off-white Beilstein Reference: 2835356 UN No.: UN2811 MDL No.: MFCD01656033

Formel: C₇H₅BrF₃N
Molecular Weight: 240,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01656033
CAS-Nummer: 58458-10-9
EINECS: 261-261-6
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR