4,6-Dichlor-o-kresol 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3,5-Dichlor-2-hydroxytoluol, 2,4-Dichlor-6-methylphenol, 1,3-Dichlor-4-hydroxy-5-methylbenzol
H64312.03
H64312.06
H64312.03EA
48.3
EUR
H64312.03
H64312.06
4,6-Dichlor-o-kresol 98%
4,6-Dichlor-o-kresol
Purity: 98%
CAS No.: 1570-65-6
Molecular Formula: C7H6Cl2O
Formula Weight: 177.02
Physical Form: Solid
Appearance: Brown
Melting Point: 52-56°
Boiling Point: 225° lit.
Beilstein Reference: 1864865
UN No.: UN3077
MDL No.: MFCD00019988
Formel:
C₇H₆Cl₂O Molecular Weight: 177.03 g/mol Siedepunkt: 225 °C (Lit.) Schmelzpunkt: 52…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019988 CAS-Nummer: 1570-65-6 EINECS: 216-382-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...