2-chloro-5-iodo-3-(trifluoromethyl)pyridine 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64329.03
H64329.06
H64329.03EA
58.3
EUR
H64329.03
H64329.06
2-chloro-5-iodo-3-(trifluoromethyl)pyridine 95%
2-chloro-5-iodo-3-(trifluoromethyl)pyridine
Purity: 95%
CAS No.: 887707-25-7
Molecular Formula: C6H2ClF3IN
Formula Weight: 307.44
Physical Form: Solid
Sensitivity: Light Sensitive
Melting Point: 43°
Beilstein Reference: 10249610
MDL No.: MFCD12024302
Formel:
C₆H₂ClF₃IN Schmelzpunkt: 43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12024302 CAS-Nummer: 887707-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...