2-Amino-4-brompyrimidin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Bromopyrimidin-2-amine
H64358.03
H64358.03EA
294
EUR
H64358.03
2-Amino-4-brompyrimidin 98%
2-Amino-4-bromopyrimidine
Purity: 98%
CAS No.: 343926-69-2
Molecular Formula: C4H4BrN3
Formula Weight: 174.00
Physical Form: Powder
Appearance: White
Beilstein Reference: 111375
MDL No.: MFCD09999168
Formel:
C₄H₄BrN₃ Molecular Weight: 174,00 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09999168 CAS-Nummer: 343926-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...