4-Bromo-3-chloro-2-fluoropyridine 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64362.03
H64362.06
H64362.03EA
95.5
EUR
H64362.03
H64362.06
4-Bromo-3-chloro-2-fluoropyridine 98%
4-Bromo-3-chloro-2-fluoropyridine
Purity: 98%
CAS No.: 1017793-21-3
Molecular Formula: C5H2BrClFN
Formula Weight: 210.43
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD11040607
Formel:
C₅H₂BrClFN |
MDL:
MFCD11040607 CAS-Nummer: 1017793-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...