5-Bromo-3-chloropyridin-2(1H)-one 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Bromo-3-chloro-2-hydroxypyridine
H64370.06
H64370.06EA
318
EUR
H64370.06
5-Bromo-3-chloropyridin-2(1H)-one 97%
5-Bromo-3-chloropyridin-2(1H)-one
Purity: 97%
CAS No.: 58236-70-7
Molecular Formula: C5H3BrClNO
Formula Weight: 208.44
Physical Form: Solid
Melting Point: 199-202°
Beilstein Reference: 1448359
MDL No.: MFCD05662389
Formel:
C₅H₃BrClNO Molecular Weight: 208,44 g/mol Schmelzpunkt: 199…202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05662389 CAS-Nummer: 58236-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...