9-(4-Bromophenyl)carbazole 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
9-(4-Bromophenyl)-9H-carbazole
H64404.06
H64404.06EA
48.1
EUR
H64404.06
9-(4-Bromophenyl)carbazole 98%
9-(4-Bromophenyl)carbazole
Purity: 98%
CAS No.: 57102-42-8
Molecular Formula: C18H12BrN
Formula Weight: 322.20
Physical Form: Solid
Melting Point: 147-151°
Beilstein Reference: 199590
MDL No.: MFCD11617969
Formel:
C₁₈H₁₂BrN Molecular Weight: 322,21 g/mol Schmelzpunkt: 147…151 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11617969 CAS-Nummer: 57102-42-8 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...