Methyl-5-bromo-2-(methylthio)pyrimidine-4-carboxylate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 5-bromo-2-(methylthio)pyrimidine-4-carboxylate
H64417.06
H64417.06EA
288
EUR
H64417.06
Methyl-5-bromo-2-(methylthio)pyrimidine-4-carboxylate 97%
Methyl-5-bromo-2-(methylthio)pyrimidine-4-carboxylate
Purity: 97%
CAS No.: 50593-91-4
Molecular Formula: C7H7BrN2O2S
Formula Weight: 263.11
Physical Form: Solid
Appearance: Pale yellow
Melting Point: 61-64°
Beilstein Reference: 910703
UN No.: UN2811
MDL No.: MFCD05664428
Formel:
C₇H₇BrN₂O₂S Molecular Weight: 263,11 g/mol Schmelzpunkt: 61…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664428 CAS-Nummer: 50593-91-4 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...