1-N-Cbz-4-aminopiperidine 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-1-piperidinecarboxylic acid Benzyl ester, 4-Amino-1-benzyloxycarbonylpiperidine, 4-Amino-1-carbobenzoxypiperidine, Benzyl 4-aminopiperidine-1-carboxylate, 4-Amino-1-Cbz-piperidine, 1-N-Cbz-4-Aminopiperidine, Benzyl 4-Amino-1-piperidinecarboxylate

H64424.06
H64424.06EA 463 EUR
H64424.06
1-N-Cbz-4-aminopiperidine 97%
1-N-Cbz-4-aminopiperidine

Purity: 97% CAS No.: 120278-07-1 Molecular Formula: C13H18N2O2 Formula Weight: 234.30 Beilstein Reference: 10003356 MDL No.: MFCD05863884

Formel: C₁₃H₁₈N₂O₂
Molecular Weight: 234,30 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863884
CAS-Nummer: 120278-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR