5-Brom-2-chlorbenzaldehyd 98%

Lieferant: Thermo Scientific
Achtung

H64438.06
H64438.06EA 241 EUR
H64438.06
5-Brom-2-chlorbenzaldehyd 98%
5-Brom-2-chlorbenzaldehyd

Purity: 98% CAS No.: 189628-37-3 Molecular Formula: C7H4BrClO Formula Weight: 219.46 Physical Form: Solid Appearance: Off-white to pale yellow Beilstein Reference: 9757388 MDL No.: MFCD08445659

Formel: C₇H₄BrClO
Molecular Weight: 219,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445659
CAS-Nummer: 189628-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR