Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, Methyl 3,4-dihydro-3-oxo-2H-1,4-benzothiazine-6-carboxylate
H64467.06
H64467.14
H64467.06EA
118
EUR
H64467.06
H64467.14
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate 97%
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Purity: 97%
CAS No.: 188614-01-9
Molecular Formula: C10H9NO3S
Formula Weight: 223.25
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 97-99°
Beilstein Reference: 14311523
MDL No.: MFCD00449104
Formel:
C₁₀H₉NO₃S Molecular Weight: 223.25 g/mol Schmelzpunkt: 97…99 °C |
MDL:
MFCD00449104 CAS-Nummer: 188614-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...