4-iodobenzene-1,2-diamine 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64469.MD
H64469.03
H64469.06
H64469.MDEA
44.3
EUR
H64469.MD
H64469.03
H64469.06
4-iodobenzene-1,2-diamine 95%
4-iodobenzene-1,2-diamine
Purity: 95%
CAS No.: 21304-38-1
Molecular Formula: C6H7IN2
Formula Weight: 234.04
Physical Form: Solid
Sensitivity: Light Sensitive
Melting Point: 73-77°
Beilstein Reference: 2716284
UN No.: UN2811
MDL No.: MFCD08669466
Formel:
C₆H₇IN₂ Schmelzpunkt: 73…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669466 CAS-Nummer: 21304-38-1 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...